missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELAREM™ Perform-FD PLUS I GMP Grade
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
Description
ELAREM™ Perform-FD PLUS GMP Grade is a fibrinogen-depleted and anticoagulant-free Human Platelet Lysate of EU origin. As a xeno-free and easy-to-use cell culture supplement. ELAREM™ Perform-FD PLUS supports the efficient and safe in vitro expansion of various primary cells and cell lines. ELAREM™ Perform-FD PLUS GMP Grade is suited to cell culture applications that are subject to regulatory requirements bu do not require viral inactivation steps.
- Due to a unique fibrinogen-depletion technique, ELAREM™ Perform-FD PLUS is a clot-free and anticoagulant-free Human Platelet Lysate. As a result, the product neither requires the addition of anticoagulants in cell culture nor during manufacturing. That makes ELAREM™ Perform-FD PLUS a ready-to-use Human Platelet Lysate that is widely applicable, since the absence of anticoagulants offers even more convenient cell growth conditions.
- ELAREM™ Perform-FD PLUS is manufactured under an ISO 9001:2015 certified Quality Management System. Each batch is produced from large pools of platelet units. That ensures batch-to-batch consistency and enables reproducible conditions. ELAREM™ Perform-FD PLUS is manufactured from platelet units obtained from healthy blood donors at licensed blood centres in the EU.
Specifications
Specifications
| Form | Liquid |
| Product Type | Cell Culture Supplement |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction