Learn More
Labcon™ PerformR™ 12x75mm Round Bottom Flow Cytometry Tubes
GREENER_CHOICE
Product Code. 30280329
Change view
Click to view available options
Color:
Blue
Green
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
30280329 Green Pack of 1000
30280332 Blue Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30280329 Supplier Labcon™ Supplier No. 33503670009
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Polypropylene tubes for cell sorting and general flow cytometry use
  • Flow Cytometry Tubes
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Centrifuging to 3,000 x G
  • Freezable to -80C
  • Autoclavable
  • Capacity: 5 mL
  • Seal with plug or dual position caps sold separately
  • 12 mm W. 75 mm H.
  • Made in USA
Specifications
Certifications/Compliance ISO 9001:2015, FDA CFR 21 GMP
Color Green
Product Type Flow Cytometer Accessory