Learn More
Labcon™ PerformR™ 12x75mm Round Bottom Flow Cytometry Tubes
GREENER_CHOICE
Product Code. p-200112170
Click to view available options
Color:
Blue
Green
Unit Size:
Pack of 1000
Product Code. 30280329
Brand: Labcon™ 33503670009
Polypropylene tubes for cell sorting and general flow cytometry use
  • Flow Cytometry Tubes
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Centrifuging to 3,000 x G
  • Freezable to -80C
  • Autoclavable
  • Capacity: 5 mL
  • Seal with plug or dual position caps sold separately
  • 12 mm W. 75 mm H.
  • Made in USA
Specifications
Certifications/Compliance ISO 9001:2015, FDA CFR 21 GMP
Color Green
Product Type Flow Cytometer Accessory