missing translation for 'onlineSavingsMsg'
Learn More

Pericyazine, TRC

Product Code. 30008761
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008761 250 mg 250mg
17954085 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008761 Supplier Toronto Research Chemicals Supplier No. TRCP287360250MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2622-26-6
Molecular Formula C21 H23 N3 O S
Molecular Weight (g/mol) 365.49
Synonym 10H-Phenothiazine-2-carbonitrile, 10-[3-(4-hydroxy-1-piperidinyl)propyl]-Phenothiazine-2-carbonitrile, 10-[3-(4-hydroxypiperidino)propyl]- (6CI,7CI,8CI), 10-[3-(4-Hydroxypiperidino)propyl]phenothiazine-2-carbonitrile, 2-Cyano-10-[3-(4-hydroxy-1-piperidyl)propyl]phenothiazine, 2-Cyano-10-[3-(4-hydroxypiperidino)propyl]phenothiazine, Aolept, Bayer 1409, FI 6145, Nelactil, Nemactil, Neulactil, Neuleptil, Periciazine, Periciazinum, Pericyazine, Properciazine, Propericiazine, Propericyazine, RP 8909, SKF 20716
IUPAC Name 10-[3-(4-hydroxypiperidin-1-yl)propyl]phenothiazine-2-carbonitrile
SMILES OC1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C#N)CC1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H23N3OS/c22-15-16-6-7-21-19(14-16)24(18-4-1-2-5-20(18)26-21)11-3-10-23-12-8-17(25)9-13-23/h1-2,4-7,14,17,25H,3,8-13H2
Formula Weight 365.1562
Percent Purity >95%
Chemical Name or Material Pericyazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.