missing translation for 'onlineSavingsMsg'
Learn More

Perindoprilat Lactam B, TRC

Product Code. 30012469
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012469 100 mg 100mg
17912565 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012469 Supplier Toronto Research Chemicals Supplier No. TRCP287575100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130061-28-8
Molecular Formula C17 H26 N2 O4
Molecular Weight (g/mol) 322.4
Synonym Pyrazino[1,2-a]indole-2(1H)-acetic acid, decahydro-3-methyl-1,4-dioxo-α-propyl-, (αS,3S,5aS,9aS,10aR)-, Pyrazino[1,2-a]indole-2(1H)-acetic acid, decahydro-3-methyl-1,4-dioxo-α-propyl-, [3S-[2(R*),3α,5aβ,9aβ,10aα]]- (9CI), Perindoprilat lactam B, (S)-2-{(3S,5aS,9aS,10aR)-3-methyl-1,4-dioxodecahydropyrazino[1,2-a]indol-2(1H)-yl}pentanoic acid, Perindopril Related Compound D (USP)
IUPAC Name (2S)-2-[(3S,5aS,9aS,10aR)-3-methyl-1,4-dioxo-5a,6,7,8,9,9a,10,10a-octahydro-3H-pyrazino[1,2-a]indol-2-yl]pentanoic acid
SMILES CCC[C@H](N1[C@@H](C)C(=O)N2[C@H]3CCCC[C@H]3C[C@@H]2C1=O)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H26N2O4/c1-3-6-13(17(22)23)18-10(2)15(20)19-12-8-5-4-7-11(12)9-14(19)16(18)21/h10-14H,3-9H2,1-2H3,(H,22,23)/t10-,11-,12-,13-,14+/m0/s1
Formula Weight 322.1893
Percent Purity >95%
Chemical Name or Material (S)-2-{(3S,5aS,9aS,10aR)-3-methyl-1,4-dioxodecahydropyrazino[1,2-a]indol-2(1H)-yl}pentanoic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.