missing translation for 'onlineSavingsMsg'
Learn More

Periodic acid, ACS, 99.0-101.0%

Product Code. 15491167
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491167 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15491167 Supplier Thermo Scientific Alfa Aesar Supplier No. 030472.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10450-60-9 | H5IO6 | 227.94 g/mol

Periodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in the oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color White
CAS 10450-60-9
Melting Point 126°C to 130°C
Sensitivity Hygroscopic; Light sensitive
Assay Percent Range 99.01-101.0%
Quantity 50 g
Molecular Formula H5IO6
Physical Form Crystals
MDL Number MFCD00011344
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
Solubility Information Soluble in water and alcohol.
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
SMILES O[I](O)(O)(O)(O)=O
IUPAC Name tetrahydroxyiodous acid
Molecular Weight (g/mol) 227.94
PubChem CID 25289
ChEBI CHEBI:29150
Formula Weight 227.94
Grade ACS Reagent
Chemical Name or Material Periodic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.