missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15518824 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15518824 100 g Wide Mouth Amber Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518824 Supplier Avantor J.T.Baker Supplier No. T14605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
SMILES O[I](O)(O)(O)(O)=O

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 100 g
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
Formula Weight 227.96
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Periodic Acid
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.