missing translation for 'onlineSavingsMsg'
Learn More

Periodic acid, for analysis

Product Code. 15269315 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15269315 100 g 100g
15259315 25 g 25g
15279315 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15269315 Supplier Thermo Scientific Acros Supplier No. 453171000
  • ONLINE EXCLUSIVE PRICE
    85.10 €
    82.60 € / 100g
    Save 2.50 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10450-60-9 | H5IO6 | 227.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
SMILES O[I](O)(O)(O)(O)=O

Specifications

Melting Point 122.0°C
Quantity 100 g
Linear Formula H5IO6
Solubility Information Solubility in water: 3 kg/l (20°C). Other solubilities: soluble in alcohol, slightly soluble in ether
Formula Weight 227.94
Grade Analytical
Chemical Name or Material Periodic Acid
compliance-icons compliance-icons
Product Identifier
  • Periodic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.