missing translation for 'onlineSavingsMsg'
Learn More

Watson-Marlow 120 Series Drive with Pumphead

Product Code. 12555316
Change view
Click to view available options
Flow Rate:
0.0001 to 36 mL/min.
0.001 to 120 mL/min.
0.001 to 170 mL/min.
0.002 to 190 mL/min.
0.003 to 54 mL/min.
Pump Head:
3-channel Pump Head
Flip-top Pump Head
Quick-change Pump Head
Single-channel Pump Head
Twin-channel Pump Head
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Pump Head Unit Size
12555316 0.003 to 54 mL/min. Quick-change Pump Head Each
12466260 0.001 to 170 mL/min. Flip-top Pump Head Each
12426260 0.001 to 120 mL/min. Single-channel Pump Head Each
12446260 0.0001 to 36 mL/min. 3-channel Pump Head Each
12436260 0.0001 to 36 mL/min. Twin-channel Pump Head Each
12476260 0.002 to 190 mL/min. Flip-top Pump Head Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12555316 Supplier Watson-Marlow Supplier No. 010.6141.R00
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

120 series pumps are small and stackable, saving valuable space in cleanroom, LAF and biosafety cabinet installations, capable of flow rates up to 190 mL/min.

  • The 120 series is the perfect pump for maximising yield in fermentation and cell culture applications. Ultra-compact and stackable for multiple feeds such as pH control, antifoam, nutrient and buffer addition, the 120 offers the ideal combination of size and technology to optimise your process. The 120 series pump is perfectly suited to single-use systems. Error-free tube change takes only seconds.
  • Biopharmaceutical processes require accurate and repeatable pumping that is easy to validate. With an exceptional 2,000:1 speed range, the 100 series pump delivers the flexibility and purity so vital to drug trials. A keypad lock secures your pumping parameters for consistent and repeatable performance.
  • Pumphead options available as standard (others available on request):

  • 120R single channel, two roller pumphead for continuous tubing
TRUSTED_SUSTAINABILITY

Specifications

Control Type Digital Variable Speed
Display Type LED
Enclosure Rating IP31
Flow Rate 0.003 to 54 mL/min.
Maximum Pressure 30 psi
Motor Speed 0.1 to 32 rpm
External Control Manual remote control or automatic control via 4-20mA or 0-10V inputs.
Model 120U/R
Pump Head Quick-change Pump Head
Voltage 90/264 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.