missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Peristaltic Pump Drive

Product Code. 30178712 Shop All Sartorius Products
Change view
Click to view available options
Flow Rate:
0.002 to 3000 mL/min.
Unit Size:
1 sheet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
30178712 0.002 to 3000 mL/min. 1 sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30178712 Supplier Sartorius Supplier No. VFAPD00011
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Peristaltic pump drive for processing samples with Vivaflow TFF cassettes and Sartobind chromatography consumables. Designed for ultrafiltration, diafiltration and purification.

  • Tool-free pump head mounting, intuitive tube retention adjustment, and easy loading.
  • Digital control with optional remote input provides consistent flow that is instantly reversible.
  • Process fluid is confined to the tubing to ensure both the sample and the pump remain clean.
  • Easy wipe-down cleaning and washable pump heads, requiring only occasional lubrication.
TRUSTED_SUSTAINABILITY

Specifications

Control Type Digital
Display Type LCD
Enclosure Rating IP31
Flow Rate 0.002 to 3000 mL/min.
Maximum Pressure 2 bar
Motor Speed 3 to 400 rpm
Reversible Yes
Filter Type Ultrafiltration
Format Cassette
Material Polycarbonate
Material (Membrane) PES
MWCO 5 kDa
Sterility Non-sterile
Dimensions (L x W x H) 230 x 170 x 132 mm
Voltage 115/230 V
Type TFF cassette
Volume (Metric) 0.5 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.