missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Peristaltic Pump Head for 1.6 mm Tubing

Product Code. 30178741 Shop All Sartorius Products
Change view
Click to view available options
Tubing Size:
1.6 mm
Unit Size:
1 sheet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size Unit Size
30178741 1.6 mm 1 sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30178741 Supplier Sartorius Supplier No. VFAPH00011
  • ONLINE EXCLUSIVE PRICE
    729.00 €
    669.28 € / 1 sheets
    Save 59.72 €
    8% Off
Estimated Shipment: 20-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Peristaltic pump head for 1.6 mm tubing, compatible with a wide range of internal diameters.

  • Simple and intuitive set up
  • Precise flow control
  • Avoid contamination
  • Low maintenance
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W x H) 85 x 60 x 82 mm
Filter Type Ultrafiltration
Format Cassette
Material Polycarbonate
Material (Membrane) PES
MWCO 5 kDa
Sterility Non-sterile
Tubing Size 1.6 mm
Type Peristaltic Pump Head, TFF cassette
Volume (Metric) 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.