missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Peristaltic Pump Head for 2.4 mm Tubing

Product Code. 30178722 Shop All Sartorius Products
Change view
Click to view available options
Tubing Size:
2.4 mm
Unit Size:
1 sheet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size Unit Size
30178722 2.4 mm 1 sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30178722 Supplier Sartorius Supplier No. VFAPH00021
  • ONLINE EXCLUSIVE PRICE
    729.00 €
    669.28 € / 1 sheets
    Save 59.72 €
    8% Off
Estimated Shipment: 02-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Peristaltic pump head for 2.4 mm tubing, compatible with a wide range of internal diameters.

  • Simple and intuitive set up
  • Precise flow control
  • Avoid contamination
  • Low maintenance
TRUSTED_SUSTAINABILITY

Specifica

Dimensions (L x W x H) 85 x 60 x 82 mm
Filter Type Ultrafiltration
Format Cassette
Material Polycarbonate
Material (Membrane) PES
MWCO 10 kDa
Sterility Non-sterile
Tubing Size 2.4 mm
Type Peristaltic Pump Head, TFF cassette
Volume (Metric) 0.5 L
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.