missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Permabond™ FFAP GC Capillary Column

Brand:  Macherey-Nagel™ 723830.10

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10033011

  • 295.00 € / Each

Estimated Shipment: 03-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

G25, G35
GC
GC Capillary Column
220°C to 240°C
10 m
1 Each
Permabond™ FFAP
0.32 mm
0.5 mm
0.35 µm
Permabond™ FFAP
Videos
SDS
Documents
Certificates
Special Offers

Special Offers