missing translation for 'onlineSavingsMsg'
Learn More

Permethrin (3:5 cis/trans mixture), TRC

Product Code. 17978699
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17978699 100 mg 100mg
30136172 1 g 1g
30135693 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17978699 Supplier Toronto Research Chemicals Supplier No. TRCP288500100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52645-53-1
Molecular Formula C21 H20 Cl2 O3
Molecular Weight (g/mol) 391.29
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (3-phenoxyphenyl)methyl ester, (3-Phenoxyphenyl)methyl 2,2-dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylate, 3-Phenoxybenzyl 2,2-dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylate, 3-Phenoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, Acticin, Adion, Ambush, Aninsen Per-30, Anomethrin N, Antiborer 3768, Aqua-Kontrol, Astro, Bansect, Bematin 987, BioKill, Butox 50, Chinetrin, Cooper, Coopex, Corsair, Damminix, Dancide PS 150, Dichlorophenothrin, Diffusil H, Dragnet, Dragnet FT, Dragon, Ecsumin, Ectiban, Efmethrin, Eliminator Ant, Flea & Tick Killer, Elimite, Eulan SPA, Everwood PC 30W, Exmin, FMC 33297, FMC 41655, ICI-PP 557, Imperator, InfectoPedicul, Insectal Plus, Insorbcid MP, Ipitox, JF 7065, Kaleait, Kavil, Kestrel, Kestrel (pesticide), Kudos, Last Call, Lyclear, MP 79, MP 79 (ester), Mitin BC, NIA 33297, NRDC 143, Nedax, Nix, Nix (insecticide), PP 557, Perigen, Perigen 500, Perigen Defense, Perigen W, Perm-UP, Permanone, Permanone 40, Permanone 80, Permanone 90, Permasect, Permectin II, Permethrin, Permethrin I, Permethrin II, Permethrine, Permitrine, Pertrin E, Piredam, Pounce, Preventol HS 75, Pulvex, Pynosect, Repel Permanone, Riapan A, Ridect Pour-On, S 3151, SBP 1513, SBP 15131TEC, Sharkesuper, Skintex MR-III, Smite EC 10, Spartan, Spartan (pesticide), Stomoxin, Stomozan, Tanalith T, Timbertreat ICP 5, WL 43479, m-Methoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, m-Phenoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate
IUPAC Name (3-phenoxyphenyl)methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
SMILES CC1(C)C(C=C(Cl)Cl)C1C(=O)OCc2cccc(Oc3ccccc3)c2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H20Cl2O3/c1-21(2)17(12-18(22)23)19(21)20(24)25-13-14-7-6-10-16(11-14)26-15-8-4-3-5-9-15/h3-12,17,19H,13H2,1-2H3
Formula Weight 390.0789
Percent Purity >95%
Chemical Name or Material Permethrin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.