missing translation for 'onlineSavingsMsg'
Learn More

Peroxyacetic acid, ca. 35wt.% sol. in diluted acetic acid, stabilized

Product Code. 10670395 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10670395 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10670395 Supplier Thermo Scientific Acros Supplier No. 257755000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxidation reagent | CAS: 79-21-0 | C2H4O3 | 76.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-21-0 , 7722-84-1 , 64-19-7
InChI Key KFSLWBXXFJQRDL-UHFFFAOYSA-N
Synonym peracetic acid, peroxyacetic acid, estosteril, acetic peroxide, peroxoacetic acid, monoperacetic acid, osbon ac, acetyl hydroperoxide, proxitane 4002, desoxon 1
PubChem CID 6585
IUPAC Name ethaneperoxoic acid
SMILES CC(=O)OO

Specifications

Melting Point -44.0°C
Density 1.1300g/mL
Boiling Point 105.0°C
Flash Point 62°C
Packaging Plastic bottle
Molecular Formula C2H4O3
Quantity 500 mL
Specific Gravity 1.13
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in sulfuric acid, freely soluble in alcohol and ether
Molecular Weight (g/mol) 76.051
Formula Weight 76.05
Chemical Name or Material Peroxyacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.