missing translation for 'onlineSavingsMsg'
Learn More

Perylene, TRC

Product Code. 30417689
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30417689 25 g
30421747 10 g
30428080 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417689 Supplier Toronto Research Chemicals Supplier No. TRCP29130025G
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198-55-0
Molecular Formula C20 H12
Molecular Weight (g/mol) 252.31
Synonym Perylene, Dibenz[de,kl]anthracene, NSC 6512, peri-Dinaphthalene, α-Perylene
IUPAC Name perylene
SMILES c1cc2cccc3c4cccc5cccc(c(c1)c23)c45

Specifications

Quantity 25 g
InChI Formula InChI=1S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12H
Formula Weight 252.0939
Chemical Name or Material Perylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.