missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Pestle

Product Code. 10695411
Change view
Click to view available options
Diameter (Metric) Outer:
24 to 28 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
10695411 24 to 28 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10695411 Supplier Bochem™ Supplier No. 8936
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Bochem™ Pestle is a 140mm long, Stainless Steel pestle for use with a Stainless Steel mortar.

  • Stainless Steel
  • For use with Stainless Steel mortar

Specifications

Diameter (Metric) Outer 24 to 28 mm
Material Stainless Steel (18/10)
For Use With (Equipment) Mortar
Length (Metric) 140mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.