Learn More
Pyrex™ Borosilicate Glass Pestle
Product Code. 13471869
Change view
Click to view available options
Length (Metric):
110mm
125mm
Finish:
Glazed
Rough
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Finish Unit Size
13471869 110mm Glazed Each
12695327 110mm Rough Each
12682022 125mm Rough Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13471869 Supplier Pyrex Supplier No. 718342R
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion. Pyrex™ Borosilicate Glass Pestle is ideal for crushing solids into powders for easier handling. Pestles are available in a rough or glazed finish. For use with Pyrex™ Mortars, sold separately.
  • Ideal for crushing solids into powders for easier handling
  • Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion
  • Available in a rough or glazed finish
  • For use with Pyrex™ Mortars (sold separately)
Specifications
Product Type Pestle
Material Borosilicate Glass
For Use With (Equipment) Pyrex™ Mortar
Length (Metric) 110mm
Finish Glazed