missing translation for 'onlineSavingsMsg'
Learn More

Nalgene Petri Dish Rack Post

Product Code. 10468612
Change view
Click to view available options
Positions:
Post
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Positions Unit Size
10468612 Post Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10468612 Supplier Nalgene Supplier No. 7159200650
  • ONLINE EXCLUSIVE PRICE
    30.20 €
    29.70 € / Pack of 4
    Save 0.50 €
    1% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Post Polycarbonate for use with 5920/21-0060 5922-0024 and 5924-0022 petri dish rack

Post Polycarbonate for use with 5920/21-0060 5922-0024 and 5924-0022 petri dish rack

  • White Polycarbonate petri dish rack post has smooth, rounded corners, finger-grip handle and Rubber feet which make carrying up to 54 plastic-round dishes (100 x 15 mm) or 42 glass-round dishes (100 x 15 mm) safe and secure
  • Petri dish rack can be decontaminated by heat sterilization
  • Simple assembly required
  • Material: Polycarbonate
  • Color: White
  • Size: 6.5 in.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color White
Material Polycarbonate
Positions Post
Sterility Sterile
For Use With (Equipment) Petri Dish Rack Post
Size 6.5 in.
Product Type Petri Dish Rack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.