Learn More
DiversifiedBiotech PetriStickers™
Product Code. 30282301
Change view
Click to view available options
Includes:
100 Square Grid
12 Sector Pie
16 Sector Pie
32 Square Grid
50 Square Grid
6 Sector Pie
70 Square Grid
8 Sector Pie
Unit Size:
Pack of 36
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
30282301 50 Square Grid Pack of 36
30282323 32 Square Grid Pack of 36
30282330 70 Square Grid Pack of 36
30282337 100 Square Grid Pack of 36
30282281 6 Sector Pie Pack of 36
30282289 8 Sector Pie Pack of 36
30282343 12 Sector Pie Pack of 36
30282342 16 Sector Pie Pack of 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 30282301 Supplier DiversifiedBiotech Supplier No. PSTK1050
  • ONLINE EXCLUSIVE PRICE
    29.70 €
    27.86 € / Pack of 36
    Save 1.84 €
    6% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Allow for petri dishes to be labeled quickly, neatly and reliably
  • PetriStickers™ allow for petri dishes to be labeled quickly, neatly and reliably
  • Made with water-resistant ink, substrate and adhesives and will remain firmly attached at temperatures from 42° to 4°C
  • Simply remove the label's protective backing and apply it to the back of a petridish
  • After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed
Specifications
Type Petri Dish Label
Includes 50 Square Grid
Diameter (Metric) 7.62 cm