missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether 100-120°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™

Product Code. 11438732 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Unit Size:
2.5 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11438732 25 L Metal Can 25 L
10725151 2.5 L Amber Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11438732 Supplier Fisher Chemical Supplier No. P/1880/25
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

C6H14, CAS Number-64742-49-0, unii-49ib0u6mld, pentane, 2-methyl, iso-hexane, 1,1-dimethylbutane, dimethylpropylmethane, 2-methylpentan, 2-methyl-pentane, 2-methyl pentane, methyl pentane, isohexane, 2.5L, 100 deg.C, Colorless, AFABGHUZZDYHJO-UHFFFAOYSA-N, 2-methylpentane, 81849, 86.178

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
SMILES CCCC(C)C

Specifications

Color Colorless
Quantity 25 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 45mbar at 20°C
Viscosity 0.7 m2/s at 25°C
Boiling Point 100°C
IUPAC Name Petroleum ether
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether, boiling range 100-120°C
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.