missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum Ether, 100-120°C, Technical, for Water Determination In Petroleum Products, Fisher Chemical
Shop All Fisher Chemical ProductsTekniske data
Tekniske data
| CAS | 8032-32-4 |
| Color | Colorless |
| Quantity | 2.5 L |
| Packaging | Plastic-coated Glass Bottle |
| Physical Form | Liquid |
| Vapor Pressure | 45mbar at 20°C |
| MDL Number | 81849 |
| Viscosity | 0.7 m2/s at 25°C |
| Synonym | p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
| Boiling Point | 100°C |
Sikkerhed og håndtering
Produktidentifikator
- Petroleum ether, boiling range 100-120°C
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?