missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, 35° to 60°C, PHOTREX™ Reagent, J.T. Baker™

Product Code. 15568564 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15568564 4 L Narrow Mouth Amber Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15568564 Supplier Avantor J.T.Baker Supplier No. 927003

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High purity solvents characterized for use in UV, visible and IR spectrophotometry

    For Spectrophotometry

Specifications

CAS 8032-32-4
Quantity 4 L
Flash Point −18.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.68kg
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide
Grade PHOTREX™ Reagent
Chemical Name or Material Petroleum Ether, 35° to 60°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.