missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether 60-80°C, Extra Pure, SLR, Fisher Chemical™

Product Code. 10131530 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Unit Size:
1 L
2.5 L
25 L
5 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10131530 1 L Amber Glass Bottle 1 L
11418722 500 mL Amber Glass Bottle 500mL
10774761 2.5 L Amber Glass Bottle 2.5 L
10784761 5 L Metal Can 5 L
10151530 25 L Metal Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10131530 Supplier Fisher Chemical Supplier No. P/1480/15
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
SMILES CCCC(C)C

Specifications

Color Colorless
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 155 hPa
Viscosity 0.52/s at 20°C
Boiling Point 60°C
IUPAC Name Petroleum ether
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether, boiling range 60-80°C
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.