missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether 60/80

Product Code. 11353207
Change view
Click to view available options
Quantity:
1000 mL
2500 mL
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353207 1000 mL 1 L
11363207 2500 mL 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11353207 Supplier Thermo Scientific Alfa Aesar Supplier No. L13734.AU
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 8032-32-4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of Ficus thoningii.

Solubility
Insoluble in water.

Notes
Flammable, may be moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8032-32-4
MDL Number MFCD00081849
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Density 0.672
Boiling Point 60°C to 80°C
Quantity 1000 mL
Flash Point −10°C (14°F)
Odor Petroleum-like
UN Number UN1268
Merck Index 14,5489
Solubility Information Insoluble in water.
Chemical Name or Material Petroleum ether 60/80
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether 60/80
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Petroleum ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.