missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether 80-100°C, Certified AR for Analysis, N-Hexane <20%, Fisher Chemical™

Product Code. p-7073382 Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Unit Size:
2.5 L
25 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10326050

Brand: Fisher Chemical P/1840/17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
SMILES CCCC(C)C

Specifications

Color Colorless
Boiling Point 78°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 200mbar at 20°C
IUPAC Name Petroleum ether
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether 80-100ºC
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.