missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, ACS Reagent Grade, Ricca Chemical

Product Code. 31750801
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750801 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 31750801 Supplier Ricca Chemical Company Supplier No. RSOP02004C
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

(Ligroin)

Specifications

CAS 8032-32-4
Acidity Pass
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation max. 0.001%
Boiling Point 35°C to 60°C
Grade ACS
Chemical Name or Material Petroleium Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.