missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether, ACS Reagent, Honeywell Riedel-de Haën™

Product Code. 15633660
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15633660

Brand: Honeywell-Riedel-de Haen 3204471L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

ACS Reagent

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Specifications

CAS 8030-30-6
CAS Min % ≥25.0000%
CAS Max % <50.0000%
Density 0.64 g/mL at 25 °C(lit.)
Boiling Point 40°C to 60°C
Flash Point −30°C
Packaging Glass Bottle
Molecular Formula C5H12
MDL Number MFCD00009498
Quantity 1 L
UN Number UN3295
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILES CCCCC
IUPAC Name pentane
Molecular Weight (g/mol) 72.15
PubChem CID 8003
ChEBI CHEBI:37830
Grade ACS Reagent
Chemical Name or Material Petroleum ether, ACS Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.