missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether, for analysis, boiling range 60-95°C

Product Code. 10529833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10529833 2.5 L Glass Bottle 2.5 L
10246723 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10529833 Supplier Thermo Scientific Acros Supplier No. 235070025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 64742-49-0 | C6H14 | 86.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
SMILES CCCC(C)C

Specifications

Melting Point -40.0°C
Color Colorless
Acidity 0.001 meq/g max.
Density 0.7000g/mL
Quantity 2.5 L
Flash Point −26°C
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.7
Viscosity 0.57 mm2/s (25°C)
Water 0.02% max. (K.F.)
Merck Index 14, 5489
Boiling Point 60.0°C to 95.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol, benzene,, chloroform, ether, carbon disulfide, carbon, tetrachloride and oils except castor oil.
IUPAC Name 2-methylpentane
Grade Analytical
Chemical Name or Material Petroleum ether, Boiling range 60° to 95°C
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Petroleum ether, boiling range 60-95°C
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.