missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether, puriss. p.a., Honeywell™ Riedel-de Haën™

Product Code. 15627070
Click to view available options
Quantity:
2.5 L
Packaging:
Stainless Steel Can
Unit Size:
25 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15627070

Brand: Honeywell Riedel-de Haën 7739925L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Puriss. p.a., bp 30-40 °C, low boiling point hydrogen treated naphtha

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 109-66-0, 78-78-4
Density 0.63g/mL at 20°C
Boiling Point 30°C to 40°C
Quantity 2.5 L
Flash Point −49°C (−56.2°F)
Packaging Stainless Steel Can
Molecular Formula C5H12
MDL Number MFCD00009498
UN Number UN1268
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILES CCCCC
IUPAC Name pentane
Molecular Weight (g/mol) 72.15
Grade puriss. p.a.
Chemical Name or Material Petroleum ether
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.