missing translation for 'onlineSavingsMsg'
Learn More

Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals

Product Code. 13229532 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 L
Packaging:
Metal drum
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13229532 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13229532 Supplier Thermo Scientific Acros Supplier No. 444360250
  • ONLINE EXCLUSIVE PRICE
    163.00 €
    158.00 € / 25 L
    Save 5.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Petroleum ether
CAS 64742-49-0, 111-65-9, 142-82-5, 108-87-2, 110-82-7, 73513-42-5
Melting Point -30.0°C
Color Colorless
Density 0.7400g/mL
Boiling Point 100.0°C to 140.0°C
Flash Point 2°C
Odor Petroleum distillates
Packaging Metal drum
Refractive Index 1.4030 to 1.4080
MDL Number MFCD00081849
Quantity 25 L
UN Number UN1268
Specific Gravity 0.74
Synonym Naphtha (petroleum)
Solubility Information Solubility in water: insoluble.
IUPAC Name Petroleum ether
Viscosity 0.76 mm2/s (25°C)
Formula Weight 112
Vapor Pressure 27 mbar at 20°C
Grade Technical
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.