missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum spirit, BAKER ANALYZED™, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
:
1 L
Specifications
Specifications
| Density | 0.65 g/cm³ (20°C) |
| Boiling Point | 40°C to 60°C (1013 hPa) |
| Quantity | 1 L |
| Flash Point | -40°C |
| Residue after Evaporation | max. 0.001% |
| MDL Number | MFCD00081849 |
| Water | max. 0.02% |
| Synonym | Petroleum Ether |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction