missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pexacerfont

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Pexacerfont is a potent and selective corticotropin-releasing factor 1 receptor (CRF1) antagonist (IC50 = 6.1 nM). Displays >150 fold selectivity for CRF1 over CRF2b. Inhibits CRF-mediated adrenocorticotropic hormone (ACTH) release from pituitary cell culture (IC50 = 129 nM). Displays anxiolytic effects in rat behavioral paradigms.
Specifications
Specifications
| Chemical Name or Material | 8-(6-Methoxy-2-methyl-3-pyridinyl)-2,7-dimethyl-N-[(1R)-1-methylpropyl]pyrazolo[1,5-a]-1,3,5-triazin-4-amine |
| CAS | 459856-18-9 |
| Quantity | 10 mg |
| Molecular Formula | C18H24N6O |
| Formula Weight | 340.43 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction