missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pexacerfont
GREENER_CHOICE

Product Code. p-7161660
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18746336

Brand: Tocris Bioscience™ 6451/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective CRF1 antagonist; anxiolytic

Pexacerfont is a potent and selective corticotropin-releasing factor 1 receptor (CRF1) antagonist (IC50 = 6.1 nM). Displays >150 fold selectivity for CRF1 over CRF2b. Inhibits CRF-mediated adrenocorticotropic hormone (ACTH) release from pituitary cell culture (IC50 = 129 nM). Displays anxiolytic effects in rat behavioral paradigms.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-(6-Methoxy-2-methyl-3-pyridinyl)-2,7-dimethyl-N-[(1R)-1-methylpropyl]pyrazolo[1,5-a]-1,3,5-triazin-4-amine
CAS 459856-18-9
Quantity 10 mg
Molecular Formula C18H24N6O
Formula Weight 340.43
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.