missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PF 04457845

Product Code. 18750519
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18750519 5 mg 5mg
18779568 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18750519 Supplier R&D Systems™ Supplier No. 6374/5
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective irreversible FAAH inhibitor

PF 04457845 is a potent and selective irreversible FAAH inhibitor (IC50 = 7.2 nM, which is selective for FAAH over a panel of other serine hydrolases. PF 04457845 exhibits efficacy in a rat inflammatory pain model, and is orally bioavailable and brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-3-Pyridazinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methylene]-1-piperidinecarboxamide
CAS 1020315-31-4
Assay Percent Range >98%
Quantity 5 mg
Product Line Tocris
Molecular Formula C23H20F3N5O2
Formula Weight Observed MW: 455.43
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.