Learn More
PF 04457845, Tocris Bioscience™
GREENER_CHOICE
Potent and selective irreversible FAAH inhibitor
Brand:  Tocris Bioscience 6374/5
Encompass
Product Code. 15913271
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
PF 04457845 is a potent and selective irreversible FAAH inhibitor (IC50 = 7.2 nM, which is selective for FAAH over a panel of other serine hydrolases. PF 04457845 exhibits efficacy in a rat inflammatory pain model, and is orally bioavailable and brain penetrant.
Specifications
Specifications
N-3-Pyridazinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methylene]-1-piperidinecarboxamide
1020315-31-4
>98%