missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PF 04620110

Product Code. 18781388
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18781388 25 mg 25mg
18778228 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18781388 Supplier R&D Systems™ Supplier No. 5863/25
  • ONLINE EXCLUSIVE PRICE
    592.38 €
    533.40 € / 25mg
    Save 58.98 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective DGAT-1 inhibitor; orally bioavailable

PF 04620110 is a potent and selective DGAT-1 inhibitor (IC50 = 19 nM). Exhibits >1000-fold selectivity for DGAT-1 over DGAT-2, and no significant activity against a range of other enzymes, ion channels and receptors. Reduces plasma triglyceride levels in a rat lipid challenge model. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-4-[4-(4-Amino-7,8-dihydro-5-oxopyrimido[5,4-f][1,4]oxazepin-6(5H)-yl)phenyl]cyclohexaneacetic acid
CAS 1109276-89-2
Assay Percent Range >98%
Quantity 25 mg
Product Line Tocris
Molecular Formula C21H24N4O4
Formula Weight Observed MW: 396.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.