missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF-06260933, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
PF-06260933 is an orally active and highly selective inhibitor of MAP4K4 with IC50s of 3.7 and 160 nM for kinase and cell, respectively.
Specifications
Specifications
| Chemical Name or Material | PF-06260933 |
| CAS | 1811510-56-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H13ClN4 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 30 mg/mL (101.10 mM; Need ultrasonic and warming) |
| SMILES | NC1=C(C2=CC=C(Cl)C=C2)C=C(C(C=N3)=CC=C3N)C=N1 |
| Molecular Weight (g/mol) | 296.75 |
| Formula Weight | 296.75 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction