missing translation for 'onlineSavingsMsg'
Learn More

PF-06409577, MedChemExpress

Product Code. 16493888
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493888 10 mg 10mg
16440257 50 mg 50mg
16432896 25 mg 25mg
16426617 5 mg 5mg
16404936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493888 Supplier Medchem Express Supplier No. HY103683
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PF-06409577 is a potent and selective allosteric activator of AMPK α1β1γ1 isoform with an EC50 of 7 nM.

PF-06409577 is a potent and selective allosteric activator of AMPK α1β1γ1 isoform with an EC50 of 7 nM.

Specifications

Chemical Name or Material PF-06409577
CAS 1467057-23-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H16ClNO3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (292.58 mM)
SMILES O=C(C1=CNC2=C1C=C(C3=CC=C(C4(O)CCC4)C=C3)C(Cl)=C2)O
Molecular Weight (g/mol) 341.79
Formula Weight 341.79
Percent Purity 98.12%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.