missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 543 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PF 543 hydrochloride is a potent and selective SphK1 inhibitor (IC50 = 2 nM; Ki = 3.6 nM). Exhibits >100-fold selectivity for Sphk1 over Sphk2. Also exhibits >5,000 fold selectivity over S1P1-5 receptors and 48 protein and lipid kinases. Attenuates proliferation and induces necrosis in human colorectal cancer cells in vitro. Suppresses HCT-116 tumor xenograft growth in mice. Also reduces sickling, hemolysis and inflammation in a transgenic mouse model of sickle cell disease.
Specifications
Specifications
| Chemical Name or Material | (2R)-1-[[(4-[[3-Methyl-5-[(phenylsulfonyl)methyl]phenoxy]methyl]phenyl]methyl]-2-pyrrolidinemethanol hydrochloride |
| CAS | 1706522-79-3 |
| Quantity | 10 mg |
| Target | Sphingosine Kinase Inhibitors |
| Molecular Formula | C27H31NO4S.HCl |
| Formula Weight | 502.07 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction