missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 543 hydrochloride
GREENER_CHOICE

Product Code. p-7110897
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18705899

Brand: Tocris Bioscience™ 5754/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and selective SphK1 inhibitor

PF 543 hydrochloride is a potent and selective SphK1 inhibitor (IC50 = 2 nM; Ki = 3.6 nM). Exhibits >100-fold selectivity for Sphk1 over Sphk2. Also exhibits >5,000 fold selectivity over S1P1-5 receptors and 48 protein and lipid kinases. Attenuates proliferation and induces necrosis in human colorectal cancer cells in vitro. Suppresses HCT-116 tumor xenograft growth in mice. Also reduces sickling, hemolysis and inflammation in a transgenic mouse model of sickle cell disease.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-1-[[(4-[[3-Methyl-5-[(phenylsulfonyl)methyl]phenoxy]methyl]phenyl]methyl]-2-pyrrolidinemethanol hydrochloride
CAS 1706522-79-3
Quantity 10 mg
Target Sphingosine Kinase Inhibitors
Molecular Formula C27H31NO4S.HCl
Formula Weight 502.07
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.