missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 573228
GREENER_CHOICE

Product Code. 18024982
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18024982 10 mg 10mg
18055522 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18024982 Supplier Tocris Bioscience™ Supplier No. 3239/10
  • ONLINE EXCLUSIVE PRICE
    326.00 €
    307.65 € / 10mg
    Save 18.35 €
    5% Off
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective FAK inhibitor

Potent and selective inhibitor of focal adhesion kinase (FAK) (IC50 = 4 nM). Displays 50 - 250-fold selectivity for FAK over other protein kinases. Blocks serum and fibronectin-directed migration and decreases focal adhesion turnover in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869288-64-2
Molecular Formula C22H20F3N5O3S

Specifications

Quantity 10 mg
Formula Weight 491.49
Purity >98%
Molecular Weight (g/mol) 491.49
Chemical Name or Material PF 573228
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.