missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ PFA Liebig Vertical Condensers

Artikelnummer. 10256971
Ansicht ändern
Click to view available options
Standard Taper Joint No:
Size 29
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Standard Taper Joint No Packungsgröße
10256971 Size 29 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10256971 Lieferant Bohlender™ Lieferanten-Nr. B 30102
  • ONLINE EXCLUSIVE PRICE
    802.90 €
    602.20 € / Pack of 1
    Save 200.70 €
    24% Off
Estimated Shipment: 04-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Providing temperature resistance from -200° to +250°C.

  • Transparent in color
  • Universal chemical resistance
  • Thin-walled cooling tube made of translucent and gastight PFA
  • Cooling jacket made of borisilicate glass
  • Hose connectors made of PP and nuts for connection of cooling water
  • Ground joint cone and socket size 29 made of PTFE
  • Distillate is only exposed to PFA/PTFE
  • Suitable for vertical assembly
  • Integrated special nut (Safe-Lab™) for easy locking and unlocking of the ground joint
  • FDA conformity

Specifications

Certifications/Compliance FDA
Description Thin-walled
Includes Hose connectors, nuts
Length (Metric) 300 mm
Standard Taper Joint No Size 29
Type Vertical Liebig Condenser
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.