missing translation for 'onlineSavingsMsg'
Learn More

GVS PFAS-free SPE Cartrige

Product Code. 32178618
Change view
Click to view available options
Bed Weight:
150 mg
200 mg
500 mg
Unit Size:
Pack of 30
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Unit Size
32178618 500 mg Pack of 30
32178140 200 mg Pack of 30
32178407 150 mg Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32178618 Supplier GVS Supplier No. SPEB00WAX06500APF
  • ONLINE EXCLUSIVE PRICE
    772.93 €
    725.00 € / Pack of 30
    Save 47.93 €
    6% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for selective extraction of acidic compounds like PFAS from environmental samples.

  • Utilizes a dual retention mechanism combining ion exchange and hydrophobic interaction for efficient extraction of PFAS and other acidic compounds.
  • Ensures high recovery and precision for both short- and long-chain PFAS, delivering consistent analytical performance.
  • Effectively reduces background noise to enhance detection sensitivity during LC-MS/MS analysis.
  • Complies with EPA Methods 533, 537.1, and 1633, supporting standardized and validated testing procedures.
  • Streamlined workflow simplifies sample preparation, ensuring reliable and reproducible results across different matrices.

Specifications

Bed Weight 500 mg
For Use With (Equipment) Vacuum Manifolds or Automated SPE Equipment
Sorbent WAX
Type Solid Phase Extraction Column
Volume (Metric) 6 mL
Pore Size 40 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.