missing translation for 'onlineSavingsMsg'
Learn More

PFAS-free Water, Waternation

Product Code. 31921348 Shop All Waternation Medical Division Products
Change view
Click to view available options
Quantity:
1 L
6 L
8 L
20 L
25 L
Unit Size:
1 L
20 L
25 L
6 L
8 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31921348 1 L 1 L
31921347 6 L 6 L
31921345 8 L 8 L
31921340 20 L 20 L
31921341 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31921348 Supplier Waternation Supplier No. WNPF10010
  • ONLINE EXCLUSIVE PRICE
    100.61 €
    70.83 € / 1 L
    Save 29.78 €
    29% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PFAS-free water is specifically designed for PFAS analysis and is compatible with ultra-trace analytical methods. It can also be used for PFAS-free production processes.

PFAS-Free Water is reagent-grade pure water with no detectable PFAS. Specifically developed for trace and ultra-trace analysis of per- and polyfluoroalkyl substances, it eliminates background interference from the water itself, so your results reflect your sample, not your solvent. PFAS Free water is designed for laboratories that demand the lowest possible detection limits, our PFAS-Free Water is trusted by:

  • Environmental monitoring laboratories
  • Drinking water and wastewater testing facilities
  • Food and beverage safety laboratories
  • Contract analytical laboratories
  • Governmental and regulatory institutions
  • PFAS testing
  • It is manufactured to ensure consistent and reliable quality. The product is available in a range of sizes and packaging options.

In PFAS analysis, if the water used contains even a small amount of PFAS contamination, it can:

  • Cause false positive results
  • Samples needing to be re-run
  • Wasted time
  • QC failures
  • Re-analysis costs that are often far higher than the cost of PFAS-free water

Chemical Identifiers

CAS 7732-18-5

Specifications

Molecular Weight (g/mol) 18.015
Percent Purity 99.99%
Grade Environmental, Extra Pure, Analytical, Purified, Technical
Packaging Bottle
Quantity 1 L
Chemical Name or Material Pure water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.