missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFTK1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
226.00 € - 468.00 €
Specifications
| Antigen | PFTK1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PFTK1 Polyclonal antibody specifically detects PFTK1 in Human samples. It is validated for Western BlotSpecifications
| PFTK1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.3), 50% glycerol | |
| 5218 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell division protein kinase 14, cyclin-dependent kinase 14, EC 2.7.11, KIAA0834EC 2.7.11.22, PFTAIRE1hPFTAIRE1, PFTK1PFTAIRE protein kinase 1, Serine/threonine-protein kinase PFTAIRE-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 312-451 of human PFTK1 (NP_036527.1). AAFPGMKDIQDQLERIFLVLGTPNEDTWPGVHSLPHFKPERFTLYSSKNLRQAWNKLSYVNHAEDLASKLLQCSPKNRLSAQAALSHEYFSDLPPRLWELTDMSSIFTVPNVRLQPEAGESMRAFGKNNSYGKSLSNSKH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title