missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PGFM Enzyme Competitive ELISA Kit

Product Code. 17825017
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Pack of 10
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17825017 10 x 96 Tests Pack of 10
15815863 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17825017 Supplier Invitrogen™ Supplier No. EIAPGFMX10
  • ONLINE EXCLUSIVE PRICE
    5280.00 €
    4804.80 € / Pack of 10
    Save 475.20 €
    9% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ELISA

The PGFM Enzyme ELISA quantitates PGFM in serum,plasma,urine,dried fecal extracts,or cell culture medium.

Principle of the method

The PGFM Competitive ELISA research-use-only kit designed to quantitatively measure PGFM present in fecal extracts,urine,serum,and plasma samples. A PGFM standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit IgG. A PGFM-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a rabbit polyclonal antibody highly specific to PGFM to each well. After a 1-hour incubation,the plate is washed and substrate is added. The substrate reacts with the bound PGFM-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

In many species, uterine and placental Prostaglandin F2alpha (PGFM) is involved in the regulation of reproductive and pregnancy-related processes such as embryonic development, initiation of parturition, and resumption of ovarian activity. Prostaglandin F2alpha is metabolized to PGFM during the first passage through the lungs. PGFM has a longer half-life in peripheral circulation than PGF2alpha and has been applied as a useful analytical marker of PGF2alpha. PGFM is identical across all species and so this kit should measure PGFM from sources other than those tested.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 50-3200 pg/mL
Assay Sensitivity 20.8 pg/mL
Description PGFM Competitive ELISA Kit
Assay Multiplex Assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Sufficient For 10 x 96 Assays
Target Species Chemical
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Prostaglandin FMProstaglandin FM;
Interassay CV 9.7%
Intraassay CV 9.2%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Protein Aliases C20H34O5; 13,14-dihydro-15-keto-PGF2alpha; 13,14-dihydro-15-keto PGF2a; 13,14-dihydro-15-keto PGF2 alpha
Quantity 10 x 96 Tests
Regulatory Status RUO
Research Discipline Reproductive Biology
Sample Volume Fecal Extract 50 μL, Plasma 6.25 μL, Serum 6.25 μL, Supernatant 50 μL, Urine 6.25 μL
Shipping Condition Wet or Dry Ice
Storage Requirements -20°C
Target PGFM
Test Time 1 hr 20 min
Total Assay Time 1 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.