missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Benchtop pH Meter with Three Point Calibration
Description
The HI2212 is a basic affordable pH benchtop meter. Operation is simplified to calibration, pH and temperature readout, and a last stored reading feature. All pH measurements are compensated for temperature effect automatically with the supplied HI7662 temperature probe. It also provides GLP (Good Laboratory Practice) capabilities.
- Simple User Interface: Operation is simple with limited features requiring only a couple of buttons
- Automatic pH Calibration: Can be performed at up to 3 points using five standard buffers (4.01, 6.86, 7.01, 9.18, and 10.01) and two custom buffers
- HI1131B pH Electrode: Supplied with glass body, double junction, refillable pH electrode with an indicating sensor made of High Temperature (HT) glass
- Temperature Compensation: Automatic or manual temperature compensation of pH measurements from -20.0 to 120.0°C
- GLP Functions: Allows for the storage and retrieval of data regarding the maintenance and status of the electrode
- Stability Indicator: An hourglass appears when there is instability in the reading and disappears once the reading has stabilized
- Separate Inputs: Connection for thermistor temperature probe and a universal BNC connector for use of a wide variety of pH electrodes
Specifications
Specifications
| Includes | HI1131B pH electrode, HI7662 temperature probe, HI76404N electrode holder, HI70004 pH 4.01 buffer solution sachet, HI70007 pH 7.01 buffer solution sachet, HI7071S electrolyte solution, HI700661 cleaning solution sachet, 12 VDC adapter, and instructions |
| Accuracy (Temperature) | ±0.2°C |
| Accuracy (pH) | ±0.01 |
| Calibration | Automatic |
| Range (pH) | -2 to 14 |
| Resolution (pH) | 0.01 |
| Calibration Points | 1, 2 or 3 with five standard buffers available (pH 4.01, 6.86, 7.01, 9.18, 10.01) |
| Temperature Range (Metric) | -20.0°C to +120.0°C |
| Temperature Range (English) | -4.0°F to +248.0°F |
| Resolution (Temperature) | 0.1°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?