missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Jenway™ pH Electrode

Product Code. 11460629
Change view
Click to view available options
pH Range:
0 to 14
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range unitSize
11460629 0 to 14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460629 Supplier Cole-Parmer™ Supplier No. 1009079

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift

Specifications

Dimensions (L x W) 120 x 12 mm
Type pH Combination Electrode
For Use With (Application) Measurement of oils, biofuels, non-aqueoussamples and Tris buffers
Material Glass
pH Range 0 to 14
Temperature (Metric) Range 0°C to 80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.