missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Jenway™ pH Electrode Cleaning Solution

Product Code. 11400489
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400489 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11400489 Supplier Cole-Parmer™ Supplier No. 1615677
  • ONLINE EXCLUSIVE PRICE
    66.00 €
    58.07 € / 500mL
    Save 7.93 €
    12% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Filling Solution Type Cleaning
Quantity 500 mL
compliance-icons
Product Identifier
  • Electrode Cleaning Solution
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.